Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168475.1 | NP_001161947.1 | 3 | 717 | ||
NM_001168476.1 | NP_001161948.1 | 2 | 517 | |||
NM_001168477.1 | NP_001161949.1 | 3 | 717 | |||
NM_025905.3 | NP_080181.2 | 3 | 717 | |||
GenBank mRNA | AK005431.1 | - | 2 | 478 | ||
AK076802.1 | - | 2 | 311 | |||
AK141754.1 | - | 3 | 703 | |||
AK165626.1 | - | 3 | 722 | |||
AK166555.1 | - | 3 | 617 | |||
AK166613.1 | - | 3 | 617 | |||
BC038179.1 | - | 3 | 688 | 4485073 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK011807.1 | - | 771 | Chr. - - [-] on Build |