Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025910.3 | NP_080186.3 | 10 | 1501 | ||
GenBank mRNA | AB177385.1 | - | 9 | 1302 | ||
AK007810.1 | - | 10 | 1471 | |||
AK013451.1 | - | 10 | 1461 | |||
AK031671.1 | - | 10 | 1435 | |||
BC023462.1 | - | 10 | 1756 | 5041558 | ||
BC025109.1 | - | 10 | 1423 | 4236994 | ||
BC058242.1 | - | 10 | 1410 | 6308890 |
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174848.3 | NP_777273.2 | 19 | 5512 | ||
GenBank mRNA | AK030282.1 | - | 10 | 2957 | ||
AK162506.1 | - | 7 | 3349 |