Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194334.2 | NP_919315.2 | 11 | 2590 | ||
GenBank mRNA | AK089175.1 | - | 11 | 2556 | ||
AK129273.1 | - | 11 | 2576 | |||
AK156788.1 | - | 11 | 2577 | |||
BC056467.1 | - | 4 | 901 | |||
BC108420.1 | - | 6 | 1195 |