Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025927.4 | NP_080203.1 | 2 | 297 | ||
GenBank mRNA | AK011163.1 | - | 2 | 166 | ||
AK011605.1 | - | 2 | 156 | |||
AK081922.1 | - | 2 | 158 | |||
AK157771.1 | - | 2 | 159 | |||
AK198473.1 | - | 2 | 135 | |||
BC020130.1 | - | 2 | 142 | 3990098 | ||
BY263659.1 | - | 2 | 297 |