Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025949.3 | NP_080225.2 | 9 | 983 | ||
GenBank mRNA | AK049349.1 | - | 7 | 734 | ||
BC054113.1 | - | 7 | 656 | 30015425 |