Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012312.1 | - | 4 | 613 | ||
AK016651.1 | - | 4 | 756 | |||
AK038578.1 | - | 4 | 578 | |||
AK044291.1 | - | 4 | 726 | |||
AK048044.1 | - | 4 | 726 | |||
AK076231.1 | - | 4 | 740 | |||
AK078139.1 | - | 4 | 724 | |||
AK078468.1 | - | 4 | 740 | |||
AK078872.1 | - | 4 | 660 | |||
AK079556.1 | - | 4 | 577 | |||
AK080238.1 | - | 4 | 622 | |||
AK161465.1 | - | 4 | 635 | |||
AK165398.1 | - | 4 | 742 | |||
BC031761.1 | - | 4 | 606 | 4235549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR759230.1 | - | 365 | Chr. - - [-] on Build |