Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012312.1 | - | 5 | 706 | ||
AK016651.1 | - | 5 | 849 | |||
AK038578.1 | - | 5 | 671 | |||
AK044291.1 | - | 5 | 819 | |||
AK048044.1 | - | 5 | 819 | |||
AK076231.1 | - | 5 | 833 | |||
AK078139.1 | - | 5 | 817 | |||
AK078468.1 | - | 5 | 833 | |||
AK078872.1 | - | 5 | 753 | |||
AK079556.1 | - | 5 | 670 | |||
AK080238.1 | - | 5 | 715 | |||
AK161465.1 | - | 5 | 728 | |||
AK165398.1 | - | 5 | 835 | |||
BC031761.1 | - | 5 | 699 | 4235549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR759230.1 | - | 272 | Chr. - - [-] on Build |