Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025951.3 | NP_080227.2 | 3 | 693 | ||
NM_028744.3 | NP_083020.2 | 3 | 624 | |||
GenBank mRNA | AF411321.1 | - | 3 | 548 | ||
AK011751.1 | - | 3 | 656 | |||
AK016754.1 | - | 3 | 590 | |||
AK035523.1 | - | 3 | 678 | |||
AK170730.1 | - | 3 | 677 | |||
AY148879.1 | - | 590 | ||||
BC062144.1 | - | 3 | 600 | 6335661 |