Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025951.3 | NP_080227.2 | 5 | 945 | ||
NM_028744.3 | NP_083020.2 | 5 | 876 | |||
GenBank mRNA | AF411321.1 | - | 5 | 800 | ||
AK011751.1 | - | 5 | 908 | |||
AK016754.1 | - | 5 | 839 | |||
AK035523.1 | - | 5 | 930 | |||
AK170730.1 | - | 5 | 929 | |||
AY148879.1 | - | 839 | ||||
BC062144.1 | - | 5 | 852 | 6335661 |