Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025951.3 | NP_080227.2 | 10 | 1533 | ||
NM_028744.3 | NP_083020.2 | 10 | 1464 | |||
GenBank mRNA | AF411321.1 | - | 10 | 1388 | ||
AK011751.1 | - | 10 | 1496 | |||
AK016754.1 | - | 10 | 1427 | |||
AK035523.1 | - | 10 | 1518 | |||
AK170730.1 | - | 10 | 1517 | |||
AY148879.1 | - | 1427 | ||||
BC010257.1 | - | 541 | ||||
BC062144.1 | - | 10 | 1440 | 6335661 |