Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025958.2 | NP_080234.2 | 15 | 3747 | ||
GenBank mRNA | AK012416.1 | - | 1 | 187 | ||
AK129186.1 | - | 15 | 3718 | |||
AK145112.1 | - | 15 | 3732 | |||
AK170420.1 | - | 6 | 1451 | |||
BC056365.1 | - | 15 | 3729 | 6824847 | ||
BC058338.1 | - | 14 | 3562 |