Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025969.4 | NP_080245.1 | 1 | 363 | ||
GenBank mRNA | AK012203.1 | - | 1 | 363 | ||
AK013281.1 | - | 1 | 276 | |||
AK017477.1 | - | 1 | 331 | |||
AK029037.1 | - | 1 | 345 | |||
AK032468.1 | - | 1 | 364 | |||
AK168105.1 | - | 1 | 274 | |||
BC034297.1 | - | 1 | 338 | 4511762 |