Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025969.4 | NP_080245.1 | 2 | 544 | ||
GenBank mRNA | AK006594.1 | - | 2 | 49 | ||
AK012203.1 | - | 2 | 544 | |||
AK013281.1 | - | 2 | 457 | |||
AK017477.1 | - | 2 | 512 | |||
AK029037.1 | - | 2 | 526 | |||
AK032468.1 | - | 2 | 545 | |||
AK168105.1 | - | 2 | 455 | |||
BC034297.1 | - | 2 | 519 | 4511762 |