Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163687.1 | NP_001157159.1 | 6 | 790 | ||
NM_025972.4 | NP_080248.2 | 6 | 796 | |||
GenBank mRNA | AB162194.1 | - | 6 | 762 | ||
AK008776.1 | - | 6 | 798 | |||
AK133485.1 | - | 6 | 781 | |||
AK133658.1 | - | 6 | 798 | |||
AK146191.1 | - | 6 | 784 | |||
BC004572.1 | - | 6 | 766 | 3586255 |