Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177552.1 | NP_001171023.1 | 5 | 1000 | ||
NM_025976.5 | NP_080252.1 | 7 | 1375 | |||
GenBank mRNA | AK013874.1 | - | 7 | 1240 | ||
AK028592.1 | - | 5 | 839 | |||
AK041839.1 | - | 6 | 1114 | |||
BC027221.1 | - | 7 | 1216 | 4196466 |