Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025990.4 | NP_080266.2 | 3 | 315 | ||
GenBank mRNA | AK009329.1 | - | 3 | 311 | ||
AK009441.1 | - | 3 | 317 | |||
AK009531.1 | - | 3 | 311 | |||
AK009562.1 | - | 3 | 311 | |||
AK009580.1 | - | 3 | 315 | |||
AK009629.1 | - | 3 | 314 | |||
AK009695.1 | - | 3 | 312 | |||
AK009803.1 | - | 3 | 313 | |||
AK009835.1 | - | 3 | 315 | |||
AK010012.1 | - | 3 | 315 | |||
AK010051.1 | - | 3 | 312 | |||
AK010115.1 | - | 3 | 311 | |||
AK010143.1 | - | 2 | 257 | |||
AK010145.1 | - | 3 | 315 | |||
AK010158.1 | - | 3 | 311 | |||
AK010159.1 | - | 3 | 314 | |||
BC115526.1 | - | 3 | 294 | 40101263 | ||
BC115527.1 | - | 3 | 291 | 40101264 |