Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114085.1 | NP_001107557.1 | 6 | 772 | ||
NM_001285503.1 | NP_001272432.1 | 6 | 772 | |||
NM_001285504.1 | NP_001272433.1 | 5 | 623 | |||
NM_023317.2 | NP_075806.2 | 6 | 772 | |||
GenBank mRNA | AF290473.1 | - | 4 | 506 | ||
AF322073.1 | - | 5 | 679 | |||
AK012830.1 | - | 6 | 702 | |||
AK012861.1 | - | 6 | 720 | |||
AK037289.1 | - | 6 | 715 | |||
AK146982.1 | - | 6 | 702 | |||
AK150826.1 | - | 6 | 762 | |||
AK166858.1 | - | 5 | 602 | |||
AK169743.1 | - | 6 | 702 | |||
AK186962.1 | - | 1 | 70 | |||
BC023267.1 | - | 6 | 751 | 3672565 |