Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026039.3 | NP_080315.1 | 3 | 316 | ||
GenBank mRNA | AK012886.1 | - | 3 | 317 | ||
AK012903.1 | - | 3 | 293 | |||
AK159696.1 | - | 3 | 319 | |||
BC068306.1 | - | 3 | 241 | 30043405 |