Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133683.3 | NP_598444.1 | 6 | 1044 | ||
GenBank mRNA | AK018383.1 | - | 6 | 992 | ||
AK034298.1 | - | 6 | 1050 | |||
AK036228.1 | - | 5 | 1121 | |||
AK042426.1 | - | 5 | 1099 | |||
AK079735.1 | - | 902 | ||||
AK136778.1 | - | 1034 | ||||
AK138931.1 | - | 6 | 1032 | |||
AK155275.1 | - | 6 | 1005 | |||
AK170143.1 | - | 6 | 1004 | |||
AK171087.1 | - | 6 | 1004 | |||
AK188976.1 | - | 1 | 55 | |||
BC016895.1 | - | 6 | 1046 | 4239007 |