Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023322.2 | NP_075811.2 | 3 | 1000 | ||
GenBank mRNA | AK012630.1 | - | 2 | 364 | ||
AK013258.1 | - | 3 | 988 | |||
AK153972.1 | - | 3 | 983 | |||
BC141244.1 | - | 3 | 926 | 9055856 | ||
U62906.1 | - | 3 | 970 |