Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810474O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 3007 | ||
NM_001289662.1 | NP_001276591.1 | 3 | 2920 | |||
NM_026054.3 | NP_080330.1 | 3 | 2938 | |||
GenBank mRNA | AK031526.1 | - | 3 | 2801 | ||
AK031989.1 | - | 4 | 2970 | |||
AK079405.1 | - | 4 | 2838 | |||
AK220403.1 | - | 3 | 2921 | |||
BC023168.1 | - | 1 | 914 | |||
BC086679.1 | - | 1 | 2358 | |||
BC139435.1 | - | 3 | 2860 | 8862457 | ||
BC139436.1 | - | 4 | 2947 | 8862458 |