Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133684.3 | NP_598445.1 | 2, 3 | 483 | ||
GenBank mRNA | AK002874.1 | - | 2, 3 | 481 | ||
AK013440.1 | - | 2, 3 | 503 | |||
AK017150.1 | - | 2, 3 | 483 | |||
AK051230.1 | - | 2, 3 | 484 | |||
AK178160.1 | - | 2, 3 | 309 | |||
BC006888.1 | - | 2, 3 | 466 | 2647757 | ||
BC042686.1 | - | 2, 3 | 487 |