Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144517.4 | NP_653100.1 | 12, 13 | 1013 | ||
GenBank mRNA | AK076128.1 | - | 12, 13 | 961 | ||
AK083084.1 | - | 12, 13 | 984 | |||
AK184122.1 | - | 5, 6 | 309 | |||
AK217135.1 | - | 5, 6 | 317 |