Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 2 | 341 | ||
GenBank mRNA | AK013554.1 | - | 2 | 293 | ||
AK028937.1 | - | 2 | 267 | |||
AK028945.1 | - | 2 | 244 | |||
AK030280.1 | - | 2 | 281 | |||
AK036108.1 | - | 2 | 265 | |||
AK134887.1 | - | 2 | 298 | |||
AY029531.1 | - | 2 | 242 | |||
BC003946.1 | - | 2 | 228 | 3487343 | ||
CK617511.1 | - | 2 | 417 |