Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026072.1 | NP_080348.1 | 13 | 1381 | ||
GenBank mRNA | AK014025.1 | - | 13 | 1373 | ||
AK075902.1 | - | 13 | 1359 | |||
AK077779.1 | - | 13 | 1324 | |||
AK166770.1 | - | 13 | 1381 | |||
BC025437.1 | - | 13 | 1181 | 5025084 |