Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205239.1 | NP_001192168.1 | 15 | 1694 | ||
NM_029922.3 | NP_084198.2 | 15 | 1634 | |||
GenBank mRNA | AK009530.1 | - | 5 | 394 | ||
AK014142.1 | - | 6 | 437 | |||
AK035349.1 | - | 15 | 1558 | |||
AK075876.1 | - | 5 | 396 | |||
AK076321.1 | - | 15 | 1558 | |||
BC031387.1 | - | 14 | 1365 | 5038718 | ||
BC052064.1 | - | 15 | 1632 | |||
BC058266.1 | - | 6 | 588 | |||
BC062096.1 | - | 14 | 1551 | 5716648 |