Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205239.1 | NP_001192168.1 | 20 | 2098 | ||
NM_029922.3 | NP_084198.2 | 20 | 2038 | |||
GenBank mRNA | AK014142.1 | - | 11 | 841 | ||
BC052064.1 | - | 20 | 2036 | |||
BC058266.1 | - | 10 | 1636 | |||
BC062096.1 | - | 19 | 1955 | 5716648 |