Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004359.2 | NP_001004359.1 | 10 | 1602 | ||
NM_001005385.1 | NP_001005385.1 | 10 | 1488 | |||
NM_026081.5 | NP_080357.4 | 11 | 1691 | |||
GenBank mRNA | AB093241.1 | - | 5 | 808 | ||
AK050919.1 | - | 7 | 1126 | |||
AK083192.1 | - | 10 | 1448 | |||
BC058970.1 | - | 1 | 445 | 6826050 | ||
BC070413.1 | - | 11 | 1639 | |||
BC085157.1 | - | 10 | 1390 | 30357108 |