Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rffl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007465.3 | NP_001007466.1 | 5, 6 | 1094 | ||
NM_001164569.1 | NP_001158041.1 | 6, 7 | 1238 | |||
NM_001164570.1 | NP_001158042.1 | 6, 7 | 1180 | |||
NM_001164571.1 | NP_001158043.1 | 4, 5 | 1013 | |||
NM_026097.3 | NP_080373.1 | 4, 5 | 1121 | |||
GenBank mRNA | AF434814.1 | - | 3, 4 | 799 | ||
AK006757.1 | - | 4, 5 | 1109 | |||
AK007189.1 | - | 4, 5 | 1109 | |||
AK015806.1 | - | 4, 5 | 1122 | |||
AK028095.1 | - | 4, 5 | 1110 | |||
AK128983.1 | - | 6, 7 | 1181 | |||
AK150213.1 | - | 5, 6 | 1027 | |||
AK150541.1 | - | 6, 7 | 1200 | |||
AK159442.1 | - | 5, 6 | 1053 | |||
AK169393.1 | - | 4, 5 | 954 | |||
BC118517.1 | - | 4, 5 | 896 | 40105713 |