Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113573.1 | NP_001107045.1 | 10 | 2092 | ||
NM_001113574.1 | NP_001107046.1 | 11 | 2143 | |||
NM_023336.4 | NP_075825.3 | 10 | 2252 | |||
GenBank mRNA | AB206708.2 | - | 10 | 1954 | ||
AF269193.1 | - | 10 | 2040 | |||
AK010739.1 | - | 3 | 676 | |||
AK014429.1 | - | 2 | 304 | |||
AK136429.1 | - | 10 | 2024 | |||
AK159827.1 | - | 10 | 2078 | |||
AK160752.1 | - | 10 | 2051 | |||
AK161164.1 | - | 9 | 3286 | |||
AK167573.1 | - | 10 | 2050 | |||
AY513269.1 | - | 9 | 1918 | |||
BC031536.1 | - | 10 | 2066 | 4015879 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK142230.1 | - | 964 | Chr. - - [-] on Build |