Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833420G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113550.1 | NP_001107022.1 | 4 | 926 | ||
NM_026127.4 | NP_080403.4 | 5 | 989 | |||
GenBank mRNA | AK008565.1 | - | 4 | 929 | ||
AK008588.1 | - | 4 | 926 | |||
AK014735.1 | - | 4 | 849 | |||
AK146515.1 | - | 4 | 917 | |||
AK149119.1 | - | 5 | 1779 | |||
AK160863.1 | - | 4 | 911 | |||
BC021327.1 | - | 5 | 969 | 5066171 | ||
BC028328.1 | - | 5 | 1758 | |||
BC030738.1 | - | 5 | 957 |