Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 4 | 584 | ||
GenBank mRNA | AK018181.1 | - | 4 | 581 | ||
AK046313.1 | - | 4 | 537 | |||
AK141611.1 | - | 4 | 585 | |||
AK220169.1 | - | 4 | 572 | |||
AY174062.1 | - | 3 | 713 | |||
BC056641.1 | - | 4 | 543 | 5097345 |