Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 17 | 2274 | ||
GenBank mRNA | AK009490.1 | - | 2 | 184 | ||
AK018181.1 | - | 17 | 2271 | |||
AK081358.1 | - | 10 | 1259 | |||
AK141611.1 | - | 17 | 2275 | |||
AK208223.1 | - | 3 | 279 | |||
AK220169.1 | - | 17 | 2262 | |||
AY174062.1 | - | 16 | 2403 | |||
BC002133.1 | - | 2 | 243 | |||
BC008137.1 | - | 13 | 1541 | |||
BC056641.1 | - | 17 | 2233 | 5097345 |