Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026141.3 | NP_080417.2 | 6 | 571 | ||
GenBank mRNA | AK011443.1 | - | 6 | 543 | ||
AK012371.1 | - | 6 | 548 | |||
AK013377.1 | - | 6 | 544 | |||
AK014406.2 | - | 544 | ||||
AK014453.1 | - | 5 | 1096 | |||
AK017370.1 | - | 6 | 571 | |||
AK167168.1 | - | 6 | 536 | |||
AK167258.1 | - | 6 | 543 | |||
BC079912.1 | - | 6 | 533 | 30661438 |