Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026144.4 | NP_080420.2 | 3, 4 | 317 | ||
GenBank mRNA | AK004993.1 | - | 3, 4 | 379 | ||
AK034277.1 | - | 3, 4 | 379 | |||
AK036929.1 | - | 3, 4 | 395 | |||
AK049991.1 | - | 3, 4 | 318 | |||
AK135087.1 | - | 3, 4 | 379 | |||
AK149138.1 | - | 3, 4 | 349 | |||
AK171207.1 | - | 3, 4 | 391 | |||
BC004011.1 | - | 3, 4 | 365 | 3493585 |