Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113554.1 | NP_001107026.1 | 7 | 1606 | ||
NM_026149.4 | NP_080425.3 | 7 | 1609 | |||
GenBank mRNA | AF521132.1 | - | 7 | 1607 | ||
AK014996.2 | - | 7 | 1609 | |||
AK077673.1 | - | 7 | 1191 | |||
AK083587.1 | - | 7 | 1191 | |||
AK167855.1 | - | 7 | 1141 | |||
AK168122.1 | - | 7 | 1141 | |||
BC031583.1 | - | 7 | 1175 | 4952090 |