Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026164.2 | NP_080440.2 | 2 | 1191 | ||
GenBank mRNA | AB044139.1 | - | 2 | 1070 | ||
AK004621.1 | - | 2 | 1158 | |||
AK029616.1 | - | 2 | 1157 | |||
AK084928.1 | - | 858 | ||||
AK145776.1 | - | 2 | 1101 | |||
AK163211.1 | - | 2 | 1159 | |||
AK168475.1 | - | 2 | 1154 | |||
BC127056.1 | - | 1 | 1051 | 40109753 |