Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286560.1 | NP_001273489.1 | 8 | 937 | ||
NM_026168.4 | NP_080444.1 | 9 | 1046 | |||
NM_026355.3 | NP_080631.1 | 9 | 1046 | |||
GenBank mRNA | AK004661.1 | - | 9 | 710 | ||
AK007498.1 | - | 9 | 695 | |||
AK008971.1 | - | 9 | 702 | |||
AK009483.1 | - | 9 | 698 | |||
AK010529.1 | - | 9 | 695 | |||
AK016275.1 | - | 9 | 1047 | |||
AK017876.1 | - | 9 | 730 | |||
AK030045.1 | - | 9 | 696 | |||
AK086727.1 | - | 9 | 698 | |||
AK135968.1 | - | 6 | 383 | |||
AK162267.1 | - | 8 | 670 | |||
AK167391.1 | - | 9 | 699 | |||
BC006895.1 | - | 9 | 664 | 2651322 | ||
BC018188.1 | - | 9 | 698 | 3711066 | ||
BC026558.1 | - | 9 | 658 | 5030661 | ||
BC064749.1 | - | 9 | 759 | 30046220 |