Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026168.4 | NP_080444.1 | 3 | 628 | ||
NM_026355.3 | NP_080631.1 | 3 | 628 | |||
GenBank mRNA | AK004661.1 | - | 3 | 292 | ||
AK007498.1 | - | 3 | 277 | |||
AK008971.1 | - | 3 | 284 | |||
AK009483.1 | - | 3 | 280 | |||
AK010529.1 | - | 3 | 277 | |||
AK014413.1 | - | 3 | 284 | |||
AK016275.1 | - | 3 | 629 | |||
AK017876.1 | - | 3 | 312 | |||
AK030045.1 | - | 3 | 278 | |||
AK086727.1 | - | 3 | 280 | |||
AK167391.1 | - | 3 | 281 | |||
AK214649.1 | - | 2 | 101 | |||
BC006895.1 | - | 3 | 246 | 2651322 | ||
BC018188.1 | - | 3 | 280 | 3711066 | ||
BC026558.1 | - | 3 | 240 | 5030661 | ||
BC064749.1 | - | 3 | 341 | 30046220 |