Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026175.5 | NP_080451.4 | 4 | 546 | ||
GenBank mRNA | AI115048.1 | - | 4 | 547 | ||
AK004763.1 | - | 4 | 529 | |||
AK028829.1 | - | 4 | 498 | |||
AK029095.1 | - | 4 | 516 | |||
AK030223.1 | - | 4 | 533 | |||
AK051703.1 | - | 4 | 546 | |||
AK160061.1 | - | 4 | 529 | |||
AK202411.1 | - | 2 | 130 | |||
AK210145.1 | - | 2 | 130 | |||
AK212717.1 | - | 2 | 130 | |||
BC029753.1 | - | 4 | 510 | 2651773 |