Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163552.1 | NP_001157024.1 | 6 | 1199 | ||
NM_001163553.1 | NP_001157025.1 | 5 | 1412 | |||
NM_026193.2 | NP_080469.2 | 5 | 1412 | |||
GenBank mRNA | AK007733.1 | - | 5 | 1301 | ||
AK165570.1 | - | 5 | 1332 | |||
BC054092.1 | - | 6 | 1210 | 30006597 | ||
BC056200.2 | - | 5 | 1300 | 30015292 |