Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026194.4 | NP_080470.2 | 10 | 1154 | ||
GenBank mRNA | AK007993.2 | - | 10 | 1099 | ||
AK013382.1 | - | 3 | 417 | |||
AK032663.1 | - | 10 | 1090 | |||
AK076206.1 | - | 10 | 1095 | |||
AK129211.1 | - | 9 | 1067 | |||
AK132697.1 | - | 10 | 1118 | |||
BC058809.1 | - | 9 | 2478 | |||
BC080203.1 | - | 10 | 1132 | |||
BC099529.1 | - | 10 | 1141 | |||
BC132195.1 | - | 10 | 1106 | 40130472 | ||
BC138153.1 | - | 10 | 1106 | 8861172 |