Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290338.1 | NP_001277267.1 | 2 | 296 | ||
NM_001290339.1 | NP_001277268.1 | 2 | 251 | |||
GenBank mRNA | AK012639.1 | - | 2 | 252 | ||
AK089032.1 | - | 2 | 294 | |||
BY771310.1 | - | 2 | 263 |