Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290338.1 | NP_001277267.1 | 6 | 1626 | ||
NM_001290339.1 | NP_001277268.1 | 4 | 1326 | |||
GenBank mRNA | AK006693.1 | - | 1 | 233 | ||
AK012639.1 | - | 4 | 1325 | |||
AK089032.1 | - | 6 | 1623 |