Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290338.1 | NP_001277267.1 | 8 | 2040 | ||
NM_001290339.1 | NP_001277268.1 | 6 | 1740 | |||
GenBank mRNA | AK006693.1 | - | 3 | 647 | ||
AK012639.1 | - | 6 | 1739 | |||
AK089032.1 | - | 8 | 2037 | |||
BC019579.1 | - | 4 | 347 | |||
CX223310.1 | - | 3 | 172 |