Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026201.3 | NP_080477.1 | 8 | 951 | ||
GenBank mRNA | AF465615.1 | - | 8 | 842 | ||
AK012111.1 | - | 8 | 842 | |||
BC010199.1 | - | 8 | 829 | |||
BC034174.1 | - | 8 | 852 | |||
BC039939.1 | - | 8 | 852 | |||
BC051052.1 | - | 8 | 853 | |||
BC060130.1 | - | 8 | 839 | |||
BC079652.1 | - | 8 | 825 | 6844310 |