Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026201.3 | NP_080477.1 | 11 | 1349 | ||
GenBank mRNA | AF465615.1 | - | 11 | 1240 | ||
AK012111.1 | - | 11 | 1240 | |||
BC010199.1 | - | 11 | 1227 | |||
BC034174.1 | - | 11 | 1250 | |||
BC039939.1 | - | 11 | 1250 | |||
BC060130.1 | - | 11 | 1237 | |||
BC079652.1 | - | 11 | 1223 | 6844310 |