Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290180.1 | NP_001277109.1 | 3 | 622 | ||
NM_001290181.1 | NP_001277110.1 | 4 | 2516 | |||
NM_024437.4 | NP_077757.2 | 4 | 776 | |||
NM_024446.5 | NP_077766.3 | 4 | 716 | |||
GenBank mRNA | AA462190.1 | - | 1 | 194 | ||
AF338424.1 | - | 4 | 736 | |||
AK004924.1 | - | 4 | 2472 | |||
AK011172.1 | - | 4 | 650 | |||
AK088191.1 | - | 3 | 624 | |||
AK218756.1 | - | 1 | 346 | |||
BC069843.1 | - | 4 | 732 | 5102473 | ||
BX634778.1 | - | 1 | 317 |