Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290676.1 | NP_001277605.1 | 2 | 2507 | ||
NM_001290678.1 | NP_001277607.1 | 2 | 2507 | |||
NM_026252.4 | NP_080528.2 | 2 | 2507 | |||
GenBank mRNA | AK173229.1 | - | 2 | 2516 | ||
AY313775.1 | - | 2 | 1237 | |||
BC079599.1 | - | 2 | 315 | |||
BC115430.1 | - | 2 | 1231 | 40094123 | ||
BC115431.1 | - | 2 | 1231 | 40094124 | ||
BC145863.1 | - | 2 | 1231 | 40131166 | ||
BC145865.1 | - | 2 | 1231 | 40131168 | ||
CF182456.1 | - | 2 | 594 |