Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026257.3 | NP_080533.1 | 14 | 1357 | ||
GenBank mRNA | AK010492.1 | - | 7 | 835 | ||
AK015720.1 | - | 14 | 1341 | |||
AK048494.1 | - | 14 | 1359 |